3-Ethoxy-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Ethoxy-4-fluorobenzeneboronic acid

16542-25G 16542-5G
76908-556EA 381.93 USD
76908-556 76908-558
3-Ethoxy-4-fluorophenylboronic acid ≥97%
3-Ethoxy-4-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD09475831
CAS Number: 900174-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR