3-Bromo-6-chloro-2-fluorotoluene ≥97%

Supplier: AOBChem USA

Warning

16424-1G 16424-25G 16424-500MG 16424-5G
76908-492EA 183.54 USD
76908-492 76908-494 76908-496 76908-498
3-Bromo-6-chloro-2-fluorotoluene ≥97%
3-Bromo-6-chloro-2-fluorotoluene
Formula: C₇H₅BrClF
Storage Temperature: Refrigerator
MDL Number: MFCD09800768
CAS Number: 943830-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR