3-Formyl-4-(trifluoromethoxy)phenyboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-Formyl-4-(trifluoromethoxy)phenyl)boronic acid

16416-25G 16416-5G
76908-484EA 595.17 USD
76908-484 76908-486
3-Formyl-4-(trifluoromethoxy)phenyboronic acid ≥97% research grade
3-Formyl-4-(trifluoromethoxy)phenyboronic acid
Formula: C₈H₆BF₃O₄
MW: 233.937 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617264
CAS Number: 1310383-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR