2,4,6-Trifluoro-3-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

16346-1G 16346-25G 16346-5G
76908-448EA 63.66 USD
76908-448 76908-450 76908-452
2,4,6-Trifluoro-3-methoxybenzoic acid ≥97%
2,4,6-Trifluoro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD06660225
CAS Number: 886499-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR