3-Bromo-4-isopropoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16233-25G 16233-5G
76908-434EA 488.02 USD
76908-434 76908-436
3-Bromo-4-isopropoxybenzaldehyde ≥97%
3-Bromo-4-isopropoxybenzaldehyde
Formula: C₁₀H₁₁BrO₂
MW: 243.097 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02629653
CAS Number: 191602-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR