3-Bromo-2-chloro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

16177-1G 16177-25G 16177-500MG 16177-5G
76908-408EA 142.17 USD
76908-408 76908-410 76908-412 76908-414
3-Bromo-2-chloro-4-iodopyridine ≥97%
3-Bromo-2-chloro-4-iodopyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClIN
Storage Temperature: Refrigerator
MDL Number: MFCD03095312
CAS Number: 884494-53-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR