(2-Ethoxy-3-fluorophenyl)boronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fluoro-2-ethoxyphenylboronic acid

16068-1G 16068-25G 16068-5G
76908-390EA 66.84 USD
76908-390 76908-392 76908-394
(2-Ethoxy-3-fluorophenyl)boronic acid ≥95%
(2-Ethoxy-3-fluorophenyl)boronic acid
Formula: C₈H₁₀BFO₃
MW: 183.97 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520053
CAS Number: 1010097-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR