3-Formyl-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16037-25G 16037-5G
76908-380EA 315.09 USD
76908-380 76908-382
3-Formyl-2-methoxyphenylboronic acid ≥97%
3-Formyl-2-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.9700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08689486
CAS Number: 480424-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR