3-Bromo-5-chlorophenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

15881-1G 15881-25G 15881-500MG 15881-5G
76908-344EA 119.89 USD
76908-344 76908-346 76908-348 76908-350
3-Bromo-5-chlorophenylboronic acid pinacol ester ≥95%
3-Bromo-5-chlorophenylboronic acid pinacol ester
Formula: C₁₂H₁₅BBrClO₂
MW: 317.414 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12405327
CAS Number: 488850-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR