3-Bromo-2-chloro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15841-1G 15841-25G 15841-5G
76908-312EA 112.46 USD
76908-312 76908-314 76908-316
3-Bromo-2-chloro-5-methylbenzaldehyde ≥97%
3-Bromo-2-chloro-5-methylbenzaldehyde
Formula: C₈H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD28369624
CAS Number: 1934517-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR