3-Chloro-5-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15749-25G 15749-5G
76908-300EA 332.07 USD
76908-300 76908-302
3-Chloro-5-(trifluoromethoxy)benzaldehyde ≥97%
3-Chloro-5-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD06660305
CAS Number: 433926-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR