3-Chloro-5-(trifluoromethoxy)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-5-(trifluoromethoxy)phenyl)boronic acid

15747-1G 15747-25G 15747-5G
76908-288EA 84.88 USD
76908-288 76908-290 76908-292
3-Chloro-5-(trifluoromethoxy)phenylboronic acid ≥97%
3-Chloro-5-(trifluoromethoxy)phenylboronic acid
Formula: C₇H₅BClF₃O₃
MW: 240.372 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856024
CAS Number: 1451393-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR