3-Bromo-5-chloro-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15736-25G 15736-5G
76908-268EA 524.09 USD
76908-268 76908-270
3-Bromo-5-chloro-4-methoxybenzaldehyde ≥97%
3-Bromo-5-chloro-4-methoxybenzaldehyde
Formula: C₈H₆BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD02629407
CAS Number: 161565-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR