4-Benzyloxy-3,5-dibromo-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15730-1G 15730-25G 15730-5G
76908-258EA 117.76 USD
76908-258 76908-260 76908-262
4-Benzyloxy-3,5-dibromo-benzaldehyde ≥97%
4-Benzyloxy-3,5-dibromo-benzaldehyde
Formula: C₁₄H₁₀Br₂O₂
MW: 370.036 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01169252
CAS Number: 249515-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR