3-Bromo-4-propoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15725-25G 15725-5G
76908-250EA 453.01 USD
76908-250 76908-252
3-Bromo-4-propoxybenzaldehyde ≥97%
3-Bromo-4-propoxybenzaldehyde
Formula: C₁₀H₁₁BrO₂
Storage Temperature: Ambient
MDL Number: MFCD07432497
CAS Number: 865138-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR