3-Acetyl-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15689-1G 15689-25G 15689-500MG 15689-5G
76908-236EA 185.66 USD
76908-236 76908-238 76908-240 76908-242
3-Acetyl-4-fluorophenylboronic acid ≥97%
3-Acetyl-4-fluorophenylboronic acid
Formula: C₈H₈BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD10696641
CAS Number: 850198-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR