4-Bromo-2,6-dichloro-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

15649-1G 15649-25G 15649-5G
76908-224EA 64.72 USD
76908-224 76908-226 76908-228
4-Bromo-2,6-dichloro-ethoxybenzene ≥97%
4-Bromo-2,6-dichloro-ethoxybenzene
Formula: C₈H₇BrCl₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11855952
CAS Number: 749932-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR