4-(4-(Trifluoromethyl)-1H-imidazol-2-yl)benzonitrile ≥95%

Supplier: AOBChem USA

Warning

156462-1G 156462-25G 156462-5G 156462-500MG
76908-210EA 109.28 USD
76908-210 76908-212 76908-214 77648-772
4-(4-(Trifluoromethyl)-1H-imidazol-2-yl)benzonitrile ≥95%
4-(4-(Trifluoromethyl)-1H-imidazol-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₁H₆F₃N₃
Storage Temperature: Ambient
MDL Number: MFCD33402469
CAS Number: 33469-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR