3,6-Dichloro-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15640-25G 15640-5G
76908-204EA 418 USD
76908-204 76908-206
3,6-Dichloro-2-fluorobenzaldehyde ≥97%
3,6-Dichloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO
MW: 193.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025381
CAS Number: 916420-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR