3,6-Dichloro-2-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15638-1G 15638-25G 15638-500MG 15638-5G
76908-192EA 231.28 USD
76908-192 76908-194 76908-196 76908-198
3,6-Dichloro-2-fluorophenylboronic acid ≥97%
3,6-Dichloro-2-fluorophenylboronic acid
Formula: C₆H₄BCl₂FO₂
MW: 208.81 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27935563
CAS Number: 2377605-67-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR