4-(Benzyloxy)-3-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15609-25G 15609-5G
76908-172EA 595.17 USD
76908-172 76908-174
4-(Benzyloxy)-3-fluorobenzaldehyde ≥97%
4-(Benzyloxy)-3-fluorobenzaldehyde
Formula: C₁₄H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD11156437
CAS Number: 175968-61-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR