3-Bromo-6-chloro-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

15587-25G 15587-5G
76908-160EA 537.88 USD
76908-160 76908-162
3-Bromo-6-chloro-2-fluorobenzaldehyde ≥97%
3-Bromo-6-chloro-2-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
Storage Temperature: Refrigerator
MDL Number: MFCD11111905
CAS Number: 886615-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR