3-Bromo-6-chloro-2-fluorobenzeneboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-6-Chloro-2-fluorophenylboronic acid

15585-1G 15585-25G 15585-5G
76908-148EA 79.57 USD
76908-148 76908-150 76908-152
3-Bromo-6-chloro-2-fluorobenzeneboronic acid ≥97%
3-Bromo-6-chloro-2-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BBrClFO₂
MW: 253.261 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044930
CAS Number: 1451393-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR