3-Bromo-2-(trifluoromethoxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

15517-1G 15517-25G 15517-500MG 15517-5G
76908-112EA 155.96 USD
76908-112 76908-114 76908-116 76908-118
3-Bromo-2-(trifluoromethoxy)benzoic acid ≥95%
3-Bromo-2-(trifluoromethoxy)benzoic acid
Formula: C₈H₄BrF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD09835356
CAS Number: 403646-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR