3-Isopropoxy-2,4,6-trifluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15470-1G 15470-25G 15470-5G
76908-094EA 79.57 USD
76908-094 76908-096 76908-098
3-Isopropoxy-2,4,6-trifluorophenylboronic acid ≥97%
3-Isopropoxy-2,4,6-trifluorophenylboronic acid
Formula: C₉H₁₀BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD07784388
CAS Number: 871125-73-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR