3-Chloro-5-(piperidine-1-carbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-5-(piperidine-1-carbonyl)phenyl)boronic acid, 3-Chloro-5-(piperidin-1-carbonyl)phenylboronic acid

15438-1G 15438-25G 15438-500MG 15438-5G
76908-070EA 138.98 USD
76908-070 76908-072 76908-074 76908-076
3-Chloro-5-(piperidine-1-carbonyl)phenylboronic acid ≥97% research grade
3-Chloro-5-(piperidine-1-carbonyl)phenylboronic acid
Formula: C₁₂H₁₅BClNO₃
MW: 267.5200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800859
CAS Number: 957120-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR