3-Chloro-5-(4-morpholinylcarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-(morpholine-4-carbonyl)phenylboronic acid

15434-1G 15434-25G 15434-500MG 15434-5G
76908-038EA 138.98 USD
76908-038 76908-040 76908-042 76908-044
3-Chloro-5-(4-morpholinylcarbonyl)phenylboronic acid ≥97% research grade
3-Chloro-5-(4-morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BClNO₄
Storage Temperature: Ambient
MDL Number: MFCD09800860
CAS Number: 957120-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR