3-Chloro-5-(diethylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-5-(diethylcarbonyl)phenylboronic acid, (3-Chloro-5-(diethylcarbamoyl)phenyl)boronic acid

15432-1G 15432-25G 15432-500MG 15432-5G
76908-022EA 138.98 USD
76908-022 76908-024 76908-026 76908-028
3-Chloro-5-(diethylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-5-(diethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₃
MW: 255.5100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800862
CAS Number: 957120-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR