3,5-Dichloro-2-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15422-1G 15422-25G 15422-500MG 15422-5G
76908-006EA 231.28 USD
76908-006 76908-008 76908-010 76908-012
3,5-Dichloro-2-methylphenylboronic acid ≥97%
3,5-Dichloro-2-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BCl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09800882
CAS Number: 957120-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR