3-(Difluoromethoxy)-4-fluorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-(Difluoromethoxy)-4-fluorophenyl)boronic acid

15413-1G 15413-25G 15413-500MG 15413-5G
76907-990EA 406.33 USD
76907-990 76907-992 76907-994 76907-996
3-(Difluoromethoxy)-4-fluorophenylboronic acid ≥97%
3-(Difluoromethoxy)-4-fluorophenylboronic acid
Formula: C₇H₆BF₃O₃
MW: 205.927 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036078
CAS Number: 958451-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR