3-Chloro-4-(methylcarbamoyl)phenylboronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
3-Chloro-4-(N-methylcarbamoyl)phenylboronic acid, (3-Chloro-4-(methylcarbamoyl)phenyl)boronic acid, 3-Chloro-4-(methylcarbamoyl)benzeneboronic acid
15383-1G
15383-25G
15383-5G
76907-982EA
62.6
USD
76907-982
76907-984
76907-986
3-Chloro-4-(methylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(methylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only
Formula:
C₈H₉BClNO₃ Storage Temperature: Ambient |
MDL Number:
MFCD07363766 CAS Number: 850589-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...