4-Benzylcarbamoyl-3-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (4-(Benzylcarbamoyl)-3-chlorophenyl)boronic acid, 3-Chloro-4-(N-benzylcarbamoyl)phenylboronic acid

15380-25G 15380-5G
76907-972EA 559.1 USD
76907-972 76907-974
4-Benzylcarbamoyl-3-chlorophenylboronic acid ≥97%
4-Benzylcarbamoyl-3-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₃BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD07363769
CAS Number: 850589-42-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR