3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-(tert-butylcarbamoyl)benzeneboronic acid, 3-Chloro-4-(N-tert-butylcarbamoyl)phenylboronic acid

15378-1G 15378-25G 15378-500MG 15378-5G
76907-956EA 124.13 USD
76907-956 76907-958 76907-960 76907-962
3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(tert-butylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₅BClNO₃
MW: 255.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07363771
CAS Number: 850589-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR