3-Chloro-4-(4-morpholinylcarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-(4-morpholinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(morpholine-4-carbonyl)phenylboronic acid

15375-1G 15375-25G 15375-500MG 15375-5G
76907-942EA 147.47 USD
76907-942 76907-944 76907-946 76907-948
3-Chloro-4-(4-morpholinylcarbonyl)phenylboronic acid ≥97% research grade
3-Chloro-4-(4-morpholinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BClNO₄
MW: 253.49 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07363774
CAS Number: 850589-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR