3-Chloro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-(1-pyrrolidinylcarbonyl)benzeneboronic acid, 3-Chloro-4-(pyrrolidinyl-1-carbonyl)phenylboronic acid

15373-1G 15373-25G 15373-500MG 15373-5G
76907-926EA 147.47 USD
76907-926 76907-928 76907-930 76907-932
3-Chloro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid ≥97% research grade
3-Chloro-4-(1-pyrrolidinylcarbonyl)phenylboronic acid
Formula: C₁₁H₁₃BClNO₃
Storage Temperature: Ambient
MDL Number: MFCD07363776
CAS Number: 850589-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR