3-Chloro-4-(cyclopentylcarbamoyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Chloro-4-cyclopentylcarbamoylphenylboronic acid, (3-Chloro-4-(cyclopentylcarbamoyl)phenyl)boronic acid

15371-1G 15371-25G 15371-5G
76907-912EA 64.72 USD
76907-912 76907-914 76907-916
3-Chloro-4-(cyclopentylcarbamoyl)phenylboronic acid ≥97%
3-Chloro-4-(cyclopentylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClNO₃
MW: 267.5200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09258664
CAS Number: 957061-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR