(3-Formyl-5-methylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Fornyl-5-methylphenylboronic acid

15363-1G 15363-25G 15363-5G
76907-894EA 62.6 USD
76907-894 76907-896 76907-898
(3-Formyl-5-methylphenyl)boronic acid ≥97%
(3-Formyl-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD08276762
CAS Number: 870777-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR