3-Fluoro-2-formyl-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

15317-1G 15317-25G 15317-500MG 15317-5G
76907-870EA 374.5 USD
76907-870 76907-872 76907-874 76907-876
3-Fluoro-2-formyl-5-methylphenylboronic acid ≥97%
3-Fluoro-2-formyl-5-methylphenylboronic acid
Formula: C₈H₈BFO₃
MW: 181.957 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28976812
CAS Number: 2121511-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR