3-Fluoro-5-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-5-methylphenyl)boronic acid, 3-Fluoro-5-methylbenzeneboronic acid

15312-25G 15312-5G
76907-862EA 332.07 USD
76907-862 76907-864
3-Fluoro-5-methylphenylboronic acid ≥97%
3-Fluoro-5-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂
Storage Temperature: Ambient
MDL Number: MFCD07363785
CAS Number: 850593-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR