3-Chloro-5-fluoro-2-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Chloro-5-fluoro-2-methoxyphenyl)boronic acid

15291-25G 15291-5G
76907-844EA 381.93 USD
76907-844 76907-846
3-Chloro-5-fluoro-2-methoxyphenylboronic acid ≥97%
3-Chloro-5-fluoro-2-methoxyphenylboronic acid
Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414927
CAS Number: 2121513-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR