3-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Ethoxy-5-trifluoromethylphenylboronic acid

15269-1G 15269-25G 15269-5G
76907-824EA 84.88 USD
76907-824 76907-826 76907-828
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥97%
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08056361
CAS Number: 871332-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR