4-Bromo-2-chloro-6-fluoroanisole ≥97%

Supplier: AOBChem USA

Warning

15224-25G 15224-5G
76907-814EA 453.01 USD
76907-814 76907-816
4-Bromo-2-chloro-6-fluoroanisole ≥97%
4-Bromo-2-chloro-6-fluoroanisole

Precaución: for Research Use Only

Formula: C₇H₅BrClFO
MW: 239.47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631477
CAS Number: 261762-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR