1,2,3-Trichloro-5-fluorobenzene ≥95% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3,4,5-Trichlorofluorobenzene

15209-1G 15209-25G 15209-5G
76907-788EA 117.76 USD
76907-788 76907-790 76907-792
1,2,3-Trichloro-5-fluorobenzene ≥95% research grade
1,2,3-Trichloro-5-fluorobenzene
Formula: C₆H₂Cl₃F
Storage Temperature: Refrigerator
MDL Number: MFCD16817656
CAS Number: 3107-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR