3-Bromo-5-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-Bromo-5-methoxyphenyl)boronic acid

15131-25G 15131-5G
76907-766EA 262.05 USD
76907-766 76907-768
3-Bromo-5-methoxyphenylboronic acid ≥95%
3-Bromo-5-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₃
MW: 230.8516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07369759
CAS Number: 849062-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR