3-Methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 3-Methoxy-5-trifluoromethylphenylboronic acid

15099-25G 15099-5G
76907-750EA 488.02 USD
76907-750 76907-752
3-Methoxy-5-(trifluoromethyl)phenylboronic acid ≥97%
3-Methoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₃
MW: 219.95 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08056362
CAS Number: 871332-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR