3-Bromo-2,5-dichlorobenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

15078-25G 15078-5G
76907-740EA 275.84 USD
76907-740 76907-742
3-Bromo-2,5-dichlorobenzotrifluoride ≥97%
3-Bromo-2,5-dichlorobenzotrifluoride
Formula: C₇H₂BrCl₂F₃
MW: 293.896 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31805643
CAS Number: 2168432-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR