3-Chloro-5-(trifluoromethyl)anisole ≥97%

Supplier: AOBChem USA

Warning

15054-1G 15054-25G 15054-5G
76907-716EA 62.6 USD
76907-716 76907-718 76907-720
3-Chloro-5-(trifluoromethyl)anisole ≥97%
3-Chloro-5-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
MW: 210.58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115855
CAS Number: 886497-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR