3-Amino-5-(methoxycarbonyl)benzeneboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Amino-5-methoxycarbonylphenylboronic acid, pinacol ester, Methyl 3-amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Methyl 3-Amino-5-boronobenzoate, pinacol ester

15013-1G 15013-25G 15013-5G
76907-706EA 93.36 USD
76907-706 76907-708 76907-710
3-Amino-5-(methoxycarbonyl)benzeneboronic acid, pinacol ester ≥97%
3-Amino-5-(methoxycarbonyl)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₄
MW: 277.124 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09258776
CAS Number: 850689-27-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR