4-Carboxy-3-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Borono-2-nitrobenzoic acid, 3-Nitro-4-carboxyphenylboronic acid, 4-Carboxy-3-nitrobenzeneboronic acid

14975-25G 14975-5G
76907-698EA 453.01 USD
76907-698 76907-700
4-Carboxy-3-nitrophenylboronic acid ≥97%
4-Carboxy-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BNO₆
Storage Temperature: Ambient
MDL Number: MFCD10696661
CAS Number: 80500-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR