(3-Cyano-4-methylphenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 3-Cyano-4-methylphenylboronic acid

14870-1G 14870-25G 14870-5G
76907-680EA 112.46 USD
76907-680 76907-682 76907-684
(3-Cyano-4-methylphenyl)boronic acid ≥97% research grade
(3-Cyano-4-methylphenyl)boronic acid
Formula: C₈H₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD10697414
CAS Number: 911210-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR